The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290368.1 | NP_001277297.1 | 13 - 14 | 1623 | 150 |
NM_001290369.1 | NP_001277298.1 | 12 - 13 | 1605 | 150 | |
NM_001290370.1 | NP_001277299.1 | 11 - 12 | 1562 | 150 | |
NM_008708.2 | NP_032734.1 | 13 - 14 | 1655 | 150 | |
GenBank mRNA | AF043327.1 | - | 13 - 14 | 1579 | 150 |
AK134521.1 | - | 13 - 14 | 1619 | 150 | |
AK161490.1 | - | 13 - 14 | 1623 | 150 | |
AK168171.1 | - | 13 - 14 | 1623 | 150 | |
AK168190.1 | - | 13 - 14 | 1623 | 150 | |
BC037647.1 | - | 12 - 13 | 1552 | 150 |