Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pip5kl1
Entrez Gene ID
227733
Gene Name
phosphatidylinositol-4-phosphate 5-kinase-like 1
Gene Aliases
BC028795
Chromosome Location
Chr.2: 32574788 - 32583780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198191.2 NP_937834.1 2 - 3 276 68
GenBank mRNA AK164998.1 - 2 - 3 379 68
AY376879.1 - 2 - 3 315 68
BC094346.1 - 2 - 3 377 68
BC117018.1 - 2 - 3 268 68
BC119037.1 - 2 - 3 268 68

View Full Product Details