Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pip5kl1
Entrez Gene ID
227733
Gene Name
phosphatidylinositol-4-phosphate 5-kinase-like 1
Gene Aliases
BC028795
Chromosome Location
Chr.2: 32574788 - 32583780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198191.2 NP_937834.1 4 - 5 490 75
GenBank mRNA AK133244.1 - 4 - 5 1898 75
AK164998.1 - 4 - 5 593 75
AY376879.1 - 4 - 5 529 75
BC094346.1 - 4 - 5 591 75
BC117018.1 - 4 - 5 482 75
BC119037.1 - 4 - 5 482 75

View Full Product Details