Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dpp7
Entrez Gene ID
83768
Gene Name
dipeptidylpeptidase 7
Gene Aliases
DPPII, Dpp2, QPP
Chromosome Location
Chr.2: 25352290 - 25357313 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031843.2 NP_114031.2 1 - 2 106 131
GenBank mRNA AF285235.1 - 1 - 2 101 131
AK028293.1 - 1 - 2 106 131
AK154193.1 - 1 - 2 112 131
BC027205.1 - 1 - 2 106 131
BC052861.1 - 1 - 2 130 131

View Full Product Details