Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Speer9-ps1
- Entrez Gene ID
-
73949
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 9, pseudogene 1
- Gene Aliases
-
4930415A14Rik, SPEER-9, Speer9-ps
- Chromosome Location
-
Chr.7: 3128146 - 3144992 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001583.3 |
|
3 - 4 |
438 |
76 |
GenBank mRNA |
BY714690.1 |
- |
3 - 4 |
439 |
76 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK015142.1 |
- |
|
439 |
|
Chr. - - [-] on Build |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Downstream |
expressed sequence AU018091 |
AU018091 |
|
9.7kb
|
Anti-sense |
Chr. - 3154658 - 3169252 on Build GRCm38 |
View Full Product Details