Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ssfa2
Entrez Gene ID
70599
Gene Name
sperm specific antigen 2
Gene Aliases
5730488C15Rik, CS-1, CS1, KRAP, SPAG13, mKIAA1927
Chromosome Location
Chr.2: 79635373 - 79672966 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080558.4 NP_542125.3 6 - 7 608 60
GenBank mRNA AB093303.1 - 6 - 7 483 60
AB120565.1 - 6 - 7 493 60
AK037213.1 - 4 - 5 318 60
AK169543.1 - 6 - 7 609 60
BC122519.1 - 6 - 7 494 60
BC141883.1 - 6 - 7 493 60
CF539296.1 - 6 - 7 424 60

View Full Product Details