The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290997.1 | NP_001277926.1 | 20 - 21 | 2411 | 80 |
NM_172537.4 | NP_766125.2 | 16 - 17 | 2568 | 80 | |
NM_199238.3 | NP_954708.1 | 16 - 17 | 2529 | 80 | |
GenBank mRNA | AB091532.1 | - | 16 - 17 | 2568 | 80 |
AB091533.1 | - | 15 - 16 | 1710 | 80 | |
AK122515.1 | - | 20 - 21 | 2414 | 80 | |
BC060680.1 | - | 16 - 17 | 1823 | 80 |