The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290997.1 | NP_001277926.1 | 14 - 15 | 1664 | 69 |
NM_001291000.1 | NP_001277929.1 | 10 - 11 | 1782 | 69 | |
NM_172537.4 | NP_766125.2 | 10 - 11 | 1782 | 69 | |
NM_199238.3 | NP_954708.1 | 10 - 11 | 1782 | 69 | |
NM_199239.3 | NP_954709.1 | 9 - 10 | 1017 | 69 | |
NM_199240.3 | NP_954710.1 | 9 - 10 | 1017 | 69 | |
NM_199241.3 | NP_954711.1 | 9 - 10 | 1017 | 69 | |
GenBank mRNA | AB091532.1 | - | 10 - 11 | 1782 | 69 |
AB091533.1 | - | 9 - 10 | 963 | 69 | |
AB091534.1 | - | 9 - 10 | 963 | 69 | |
AB091535.1 | - | 9 - 10 | 963 | 69 | |
AB091536.1 | - | 9 - 10 | 963 | 69 | |
AK031307.1 | - | 10 - 11 | 1478 | 69 | |
AK052232.1 | - | 10 - 11 | 1190 | 69 | |
AK084922.1 | - | 10 - 11 | 1479 | 69 | |
AK122515.1 | - | 14 - 15 | 1667 | 69 | |
BC060680.1 | - | 10 - 11 | 1076 | 69 |