Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ell3
Entrez Gene ID
269344
Gene Name
elongation factor RNA polymerase II-like 3
Gene Aliases
A930015D22Rik
Chromosome Location
Chr.2: 121439025 - 121444278 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145973.2 NP_666085.2 1 - 2 207 64
GenBank mRNA AK139676.1 - 2 - 3 633 64
BC045151.1 - 1 - 2 207 64

View Full Product Details