Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ano5
Entrez Gene ID
233246
Gene Name
anoctamin 5
Gene Aliases
9330162L24, Gdd1, Tmem16e
Chromosome Location
Chr.7: 51510875 - 51600693 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271879.1 NP_001258808.1 8 - 9 1013 128
NM_177694.6 NP_808362.2 9 - 10 1055 128
NR_073508.1 9 - 10 1048 128
GenBank mRNA AB125740.1 - 9 - 10 1055 128
AB206762.1 - 9 - 10 1055 128
AK034197.1 - 9 - 10 1037 128
BC109163.2 - 8 - 9 935 128

View Full Product Details