The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080853.3 | NP_543129.3 | 11 - 12 | 2221 | 80 |
GenBank mRNA | AF324864.1 | - | 11 - 12 | 2054 | 80 |
AK045409.1 | - | 11 - 12 | 1984 | 80 | |
BC038375.1 | - | 11 - 12 | 1600 | 80 |