The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081131.2 | NP_001074600.1 | 6 - 7 | 1284 | 101 |
GenBank mRNA | AK049272.1 | - | - | 1275 | 101 |
AK173223.1 | - | 1 - 2 | 342 | 101 | |
BC117994.1 | - | - | 1235 | 101 |