Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptpn21
Entrez Gene ID
24000
Gene Name
protein tyrosine phosphatase, non-receptor type 21
Gene Aliases
PTPD1, PTPRL10
Chromosome Location
Chr.12: 98676741 - 98737618 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146199.1 NP_001139671.1 10 - 11 1151 75
NM_011877.2 NP_036007.2 10 - 11 1286 75
GenBank mRNA BC014691.1 - 10 - 11 1416 75
BC066856.1 - 9 - 10 1089 75
BC098500.1 - 10 - 11 1151 75

View Full Product Details