The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290994.1 | NP_001277923.1 | 9 - 9 | 4142 | 161 |
NM_021323.3 | NP_067298.2 | 8 - 8 | 4078 | 161 | |
GenBank mRNA | AK038825.1 | - | 1 - 1 | 326 | 161 |
EU233992.1 | - | 5 - 5 | 3419 | 161 |