The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145360.2 | NP_663335.2 | 3 - 4 | 659 | 90 |
GenBank mRNA | AK029302.1 | - | 3 - 4 | 292 | 90 |
AK084208.1 | - | 3 - 4 | 289 | 91 | |
AK132073.1 | - | 3 - 4 | 91 | ||
AK160832.1 | - | 3 - 4 | 293 | 90 | |
BC004801.1 | - | 2 - 3 | 280 | 90 | |
BC110313.1 | - | 3 - 4 | 297 | 90 | |
CT010310.1 | - | 2 - 3 | 237 | 90 |