The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194335.2 | NP_919316.1 | 1 - 2 | 2495 | 78 |
GenBank mRNA | AK029812.1 | - | 1 - 2 | 2496 | 78 |
BC057613.1 | - | - | 751 | 78 |