Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shisa6
Entrez Gene ID
380702
Gene Name
shisa family member 6
Gene Aliases
Gm879
Chromosome Location
Chr.11: 66211725 - 66526126 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034874.3 NP_001030046.1 3 - 4 1037 63
GenBank mRNA AK147504.1 - 3 - 4 1038 63

View Full Product Details