The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029629.2 | NP_083905.1 | 5 - 6 | 842 | 70 |
GenBank mRNA | AI841210.1 | - | 3 - 4 | 70 | |
AK002291.1 | - | 5 - 6 | 820 | 70 | |
AK005134.1 | - | 4 - 5 | 1079 | 70 | |
AK080868.1 | - | 5 - 6 | 828 | 70 | |
AK170875.1 | - | 5 - 6 | 816 | 70 | |
BC030182.1 | - | 5 - 6 | 793 | 70 |