The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172203.2 | NP_757340.1 | 4 - 5 | 476 | 79 |
GenBank mRNA | AF539799.1 | - | 4 - 5 | 330 | 79 |
AK136432.1 | - | 4 - 5 | 391 | 79 | |
AY174116.1 | - | 1 - 2 | 71 | 79 |