The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010078.3 | NP_034208.2 | 20 - 21 | 2628 | 83 |
GenBank mRNA | AK081426.1 | - | 20 - 21 | 2572 | 83 |
AK158102.1 | - | 19 - 20 | 2430 | 83 | |
BC060605.1 | - | 18 - 19 | 3207 | 83 | |
BC125345.1 | - | 20 - 21 | 2488 | 83 | |
BC125347.1 | - | 20 - 21 | 2488 | 83 | |
U43520.1 | - | 10 - 11 | 1234 | 83 |