Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Drp2
Entrez Gene ID
13497
Gene Name
dystrophin related protein 2
Gene Aliases
AW495265, DRP-2
Chromosome Location
Chr.X: 134404628 - 134456574 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010078.3 NP_034208.2 7 - 8 1208 66
GenBank mRNA AK034383.1 - 7 - 8 1151 66
AK081426.1 - 7 - 8 1152 66
AK158102.1 - 6 - 7 1010 66
BC060605.1 - 6 - 7 979 66
BC125345.1 - 7 - 8 1068 66
BC125347.1 - 7 - 8 1068 66

View Full Product Details