Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cldn16
Entrez Gene ID
114141
Gene Name
claudin 16
Gene Aliases
PCLN1, claudin-16
Chromosome Location
Chr.16: 26463135 - 26482765 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053241.5 NP_444471.1 3 - 4 734 61
GenBank mRNA AB009016.1 - 3 - 4 396 61
AF323748.1 - 3 - 4 524 61
AK085268.1 - 3 - 4 735 61
AK085333.1 - 3 - 4 710 61
BC115574.2 - 4 - 5 513 61
BC115575.2 - 4 - 5 513 61

View Full Product Details