Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trap1
Entrez Gene ID
68015
Gene Name
TNF receptor-associated protein 1
Gene Aliases
2410002K23Rik, HSP75
Chromosome Location
Chr.16: 4039977 - 4077810 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026508.2 NP_080784.1 11 - 12 1271 145
GenBank mRNA AK002409.1 - 11 - 12 1257 145
AK010341.1 - 11 - 12 1271 145
AK088471.1 - 11 - 12 1260 145
AK143494.1 - 11 - 12 1289 145
AK162067.1 - 11 - 12 1259 145
AK167181.1 - 11 - 12 1260 145
BC006685.1 - 11 - 12 1239 145
BC006864.1 - 11 - 12 1239 145
BC022912.1 - 11 - 12 1268 145

View Full Product Details