The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008762.2 | NP_032788.2 | 1 - 1 | 912 | 88 |
GenBank mRNA | BC100680.1 | - | 1 - 1 | 911 | 88 |
BC100685.1 | - | 1 - 1 | 911 | 88 | |
BC115838.1 | - | 1 - 1 | 911 | 88 |