The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033238.1 | NP_001028410.1 | 2 - 3 | 319 | 71 |
GenBank mRNA | AK045005.1 | - | 2 - 3 | 224 | 71 |
AK147367.1 | - | 2 - 3 | 304 | 71 | |
AK156295.1 | - | 3 - 4 | 350 | 71 | |
AK170640.1 | - | 2 - 3 | 264 | 71 | |
AK171350.1 | - | 2 - 3 | 264 | 71 | |
BC150934.1 | - | 2 - 3 | 275 | 71 | |
BC150938.1 | - | 2 - 3 | 275 | 71 | |
CJ110973.1 | - | 2 - 3 | 304 | 71 |