The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029457.3 | NP_083733.1 | 2 - 3 | 316 | 99 |
GenBank mRNA | AK015987.1 | - | 2 - 3 | 295 | 99 |
AK031030.1 | - | 2 - 3 | 301 | 99 | |
BC031652.1 | - | 2 - 3 | 291 | 99 | |
CJ169147.1 | - | 2 - 3 | 316 | 99 |