The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199308.2 | NP_955012.2 | 2 - 3 | 300 | 136 |
GenBank mRNA | AK140821.1 | - | 2 - 3 | 305 | 136 |
AK155568.1 | - | 3 - 4 | 272 | 136 |