Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc14
Entrez Gene ID
239839
Gene Name
coiled-coil domain containing 14
Gene Aliases
G630039H03Rik
Chromosome Location
Chr.16: 34690567 - 34725204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172824.3 NP_766412.2 11 - 12 1793 77
GenBank mRNA BC031204.1 - 11 - 12 1686 77
BC095999.1 - 11 - 12 1689 77
DV661299.1 - 4 - 5 579 77

View Full Product Details