Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc14
Entrez Gene ID
239839
Gene Name
coiled-coil domain containing 14
Gene Aliases
G630039H03Rik
Chromosome Location
Chr.16: 34690567 - 34725204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172824.3 NP_766412.2 8 - 9 1362 61
GenBank mRNA AK029725.1 - 7 - 8 2494 61
AK090294.1 - 8 - 9 1269 61
BC031204.1 - 8 - 9 1255 61
BC095999.1 - 8 - 9 1258 61
DV661299.1 - 1 - 2 148 61

View Full Product Details