The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110017.1 | NP_001103487.1 | 27 - 28 | 3410 | 62 |
NM_027341.2 | NP_081617.1 | 26 - 27 | 2792 | 62 | |
GenBank mRNA | AK122344.1 | - | 4 - 5 | 1953 | 62 |
BC117953.1 | - | 26 - 27 | 2615 | 62 | |
BC117954.1 | - | 26 - 27 | 2621 | 62 |