Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atg3
Entrez Gene ID
67841
Gene Name
autophagy related 3
Gene Aliases
2610016C12Rik, APG3, Apg3l, Atg3l, PC3-96
Chromosome Location
Chr.16: 45158829 - 45188538 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026402.3 NP_080678.1 9 - 10 934 116
GenBank mRNA AB079386.1 - 9 - 10 663 116
AK005266.1 - 9 - 10 941 116
AK011409.1 - 9 - 10 935 116
AK078877.1 - 9 - 10 927 116
AK150914.1 - 9 - 10 935 116
AK159231.1 - 9 - 10 935 116
AK213960.1 - 3 - 4 206 116
BC010809.1 - 9 - 10 932 116
BC030053.1 - 7 - 8 1656 116

View Full Product Details