Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl6
Entrez Gene ID
56297
Gene Name
ADP-ribosylation factor-like 6
Gene Aliases
1110018H24Rik, 2210411E14Rik, BBS3
Chromosome Location
Chr.16: 59613321 - 59639405 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019665.3 NP_062639.3 2 - 3 561 144
GenBank mRNA AB232697.1 - 1 - 2 124 144
AF031903.1 - 2 - 3 279 144
AK003778.1 - 2 - 3 389 144
AK152980.1 - 2 - 3 365 144
AK158832.1 - 2 - 3 614 144
AK160666.1 - 2 - 3 380 144
BC018497.1 - 2 - 3 381 144

View Full Product Details