The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019665.3 | NP_062639.3 | 4 - 5 | 695 | 96 |
GenBank mRNA | AB232697.1 | - | 3 - 4 | 258 | 96 |
AF031903.1 | - | 4 - 5 | 413 | 96 | |
AK003778.1 | - | 4 - 5 | 523 | 96 | |
AK152980.1 | - | 4 - 5 | 499 | 96 | |
AK158832.1 | - | 4 - 5 | 748 | 96 | |
AK160666.1 | - | 4 - 5 | 514 | 96 | |
BC018497.1 | - | 4 - 5 | 515 | 96 |