Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgap31
Entrez Gene ID
12549
Gene Name
Rho GTPase activating protein 31
Gene Aliases
5830477L08Rik, AU041750, Cdgap, D330026I07Rik
Chromosome Location
Chr.16: 38598343 - 38713035 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020260.2 NP_064656.2 11 - 12 2251 129
GenBank mRNA AF151363.1 - 11 - 12 1892 129
AK129309.1 - 2 - 3 676 129
AK149975.1 - 11 - 12 2087 129
AK150109.1 - 11 - 12 2087 129
AK155729.1 - 11 - 12 2071 129
AK170685.1 - 11 - 12 2071 129
BC138870.1 - 11 - 12 2056 129

View Full Product Details