Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nit2
Entrez Gene ID
52633
Gene Name
nitrilase family, member 2
Gene Aliases
1190017B19Rik, D16Ertd502e
Chromosome Location
Chr.16: 57156665 - 57167332 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023175.1 NP_075664.1 1 - 2 54 91
GenBank mRNA AF284573.1 - 1 - 2 54 91
AK004533.1 - 1 - 2 38 91
AK004535.1 - 1 - 2 40 91
BC020153.1 - 1 - 2 43 91

View Full Product Details