The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039521.1 | NP_001034610.1 | 17 - 18 | 1847 | 95 |
GenBank mRNA | AK165676.1 | - | 17 - 18 | 1835 | 95 |
AK170340.1 | - | 2 - 3 | 162 | 95 | |
BC034110.1 | - | 13 - 14 | 1362 | 95 | |
BC138774.1 | - | 17 - 18 | 1812 | 95 | |
BC138776.1 | - | 17 - 18 | 1812 | 95 |