Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rrn3
Entrez Gene ID
106298
Gene Name
RRN3 RNA polymerase I transcription factor homolog (yeast)
Gene Aliases
AL023001, E130302O19Rik, R75565, TIF-1A, Tif1a
Chromosome Location
Chr.16: 13780699 - 13814841 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039521.1 NP_001034610.1 2 - 3 251 78
GenBank mRNA AK080397.1 - 2 - 3 206 78
AK080680.1 - - 232 78
AK165676.1 - 2 - 3 240 78
BC138774.1 - 2 - 3 216 78
BC138776.1 - 2 - 3 216 78
CJ175904.1 - 2 - 3 254 78

View Full Product Details