Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc19
Entrez Gene ID
245308
Gene Name
zinc finger, DHHC domain containing 19
Gene Aliases
Gm1744, Gm616
Chromosome Location
Chr.16: 32496241 - 32507214 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199309.2 NP_955013.1 1 - 2 39 71
GenBank mRNA AK132780.1 - 1 - 2 56 71
AK133398.1 - 1 - 2 40 71
BC049761.1 - 1 - 2 39 71

View Full Product Details