Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dirc2
Entrez Gene ID
224132
Gene Name
disrupted in renal carcinoma 2 (human)
Gene Aliases
RCC4
Chromosome Location
Chr.16: 35694903 - 35769356 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153550.3 NP_705778.1 3 - 4 819 76
GenBank mRNA AK034214.1 - 3 - 4 819 76
AK172331.1 - 3 - 4 834 76
BC037460.1 - 3 - 4 793 76

View Full Product Details