Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tdrd7
Entrez Gene ID
100121
Gene Name
tudor domain containing 7
Gene Aliases
5730495N10Rik, AI447470, PCTAIRE2BP
Chromosome Location
Chr.4: 45965027 - 46034766 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290475.1 NP_001277404.1 11 - 12 2193 98
NM_146142.2 NP_666254.1 11 - 12 2191 98
GenBank mRNA AK077641.1 - 11 - 12 2200 98
BC029689.1 - 10 - 11 2065 98

View Full Product Details