The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153054.2 | NP_694694.1 | 3 - 4 | 586 | 102 |
GenBank mRNA | AK049514.1 | - | 1 - 2 | 472 | 102 |
AK076433.1 | - | 2 - 3 | 310 | 102 | |
AK144034.1 | - | 2 - 3 | 310 | 102 | |
AY779336.1 | - | 1 - 2 | 244 | 102 | |
BC027192.1 | - | 3 - 4 | 524 | 102 | |
BC031146.1 | - | 3 - 4 | 554 | 102 |