Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tex10
Entrez Gene ID
269536
Gene Name
testis expressed gene 10
Gene Aliases
2610206N19Rik, 2810462N03Rik, BC006867
Chromosome Location
Chr.4: 48430858 - 48473464 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK141288.1 - 1 - 2 167 136
BC151176.1 - 1 - 2 139 136
BC151177.1 - 1 - 2 139 136

View Full Product Details