The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163734.1 | NP_001157206.1 | 9 - 10 | 1077 | 81 |
NM_028244.2 | NP_082520.2 | 12 - 13 | 1393 | 81 | |
GenBank mRNA | AK011155.1 | - | 5 - 6 | 319 | 81 |
AK129078.1 | - | 9 - 10 | 1077 | 81 | |
BC016569.1 | - | 12 - 13 | 1393 | 81 |