The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130876.3 | NP_570946.2 | 1 - 1 | 375 | 152 |
GenBank mRNA | AA930779.1 | - | 1 - 1 | 67 | 152 |
AF345292.1 | - | 1 - 1 | 332 | 152 |