Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Suv39h1
Entrez Gene ID
20937
Gene Name
suppressor of variegation 3-9 homolog 1 (Drosophila)
Gene Aliases
AI852103, AL022883, DXHXS7466e, H3-K9-HMTase 1, KMT1A, mIS6
Chromosome Location
Chr.X: 8061171 - 8074760 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290716.1 NP_001277645.1 2 - 3 343 104
NM_011514.2 NP_035644.1 2 - 3 362 104
GenBank mRNA AF019969.1 - 2 - 3 188 104
AF193861.1 - 2 - 3 184 104
AF193862.1 - 2 - 3 287 104
AK021174.1 - 2 - 3 344 104
AK088405.1 - 2 - 3 179 104
AK132238.1 - 2 - 3 224 104
AK139757.1 - 2 - 3 331 104
AK169389.1 - 2 - 3 170 104
AK169592.1 - 2 - 3 332 104
BC023860.1 - 2 - 3 239 104
BU053459.1 - 2 - 3 362 104

View Full Product Details