Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smr2
Entrez Gene ID
20600
Gene Name
submaxillary gland androgen regulated protein 2
Gene Aliases
MSG2, Vcs2
Chromosome Location
Chr.5: 88086556 - 88109053 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021289.2 NP_067264.2 2 - 3 136 120
GenBank mRNA U82377.1 - 2 - 3 136 120

View Full Product Details