Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rufy3
Entrez Gene ID
52822
Gene Name
RUN and FYVE domain containing 3
Gene Aliases
2810428M05Rik, 6330416M07Rik, AW455998, AW538594, D5Bwg0860e, Ripx, Rpipx
Chromosome Location
Chr.5: 88564780 - 88651417 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289774.1 NP_001276703.1 5 - 6 952 125
NM_001289775.1 NP_001276704.1 5 - 6 952 125
NM_001289776.1 NP_001276705.1 6 - 7 1374 125
NM_001289777.1 NP_001276706.1 7 - 8 1199 125
NM_027530.3 NP_081806.1 5 - 6 1320 125
GenBank mRNA AK013185.1 - 7 - 8 1200 125
AK018184.1 - 5 - 6 1298 125
AK083242.1 - 5 - 6 1255 125
AK148795.1 - 5 - 6 616 125
AK160820.1 - 5 - 6 953 125
BC017648.1 - 5 - 6 709 125
BC049127.1 - 5 - 6 1259 125
BC058259.1 - 6 - 7 1093 125
CT010404.1 - 5 - 6 693 125

View Full Product Details