Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pde1c
Entrez Gene ID
18575
Gene Name
phosphodiesterase 1C
Gene Aliases
-
Chromosome Location
Chr.6: 56069798 - 56652516 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025568.2 NP_001020739.1 12 - 13 1829 101
NM_001159952.1 NP_001153424.1 11 - 12 1802 101
NM_001159953.1 NP_001153425.1 12 - 13 1829 101
NM_001159955.1 NP_001153427.1 13 - 14 1391 101
NM_001159956.1 NP_001153428.1 11 - 12 1248 101
NM_001159957.1 NP_001153429.1 12 - 13 1275 101
NM_001159960.1 NP_001153432.1 13 - 14 1703 101
NM_011054.4 NP_035184.1 13 - 14 1806 101
GenBank mRNA AK014887.1 - 11 - 12 1219 101
AK015375.1 - 13 - 14 1360 101
AK029531.1 - 12 - 13 1273 101
AK030423.1 - 12 - 13 1831 101
AK050499.1 - 13 - 14 1704 101
AK077123.1 - 11 - 12 1197 101
AK082180.1 - 12 - 13 1816 101
AK082497.1 - 12 - 13 1614 101
AK133243.1 - 13 - 14 1375 101
AK139923.1 - 11 - 12 1776 101
BC032277.1 - 12 - 13 1758 101
L76944.1 - 12 - 13 1459 101
L76946.1 - 13 - 14 1805 101
L76947.1 - 12 - 13 1292 101

View Full Product Details