The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025568.2 | NP_001020739.1 | 8 - 9 | 1398 | 109 |
NM_001159952.1 | NP_001153424.1 | 7 - 8 | 1371 | 109 | |
NM_001159953.1 | NP_001153425.1 | 8 - 9 | 1398 | 109 | |
NM_001159955.1 | NP_001153427.1 | 9 - 10 | 960 | 109 | |
NM_001159956.1 | NP_001153428.1 | 7 - 8 | 817 | 109 | |
NM_001159957.1 | NP_001153429.1 | 8 - 9 | 844 | 109 | |
NM_001159960.1 | NP_001153432.1 | 9 - 10 | 1272 | 109 | |
NM_011054.4 | NP_035184.1 | 9 - 10 | 1375 | 109 | |
GenBank mRNA | AK014887.1 | - | 7 - 8 | 788 | 109 |
AK015375.1 | - | 9 - 10 | 929 | 109 | |
AK029531.1 | - | 8 - 9 | 842 | 109 | |
AK030423.1 | - | 8 - 9 | 1400 | 109 | |
AK050499.1 | - | 9 - 10 | 1273 | 109 | |
AK077123.1 | - | 7 - 8 | 766 | 109 | |
AK082180.1 | - | 8 - 9 | 1385 | 109 | |
AK082497.1 | - | 8 - 9 | 1183 | 109 | |
AK133243.1 | - | 9 - 10 | 944 | 109 | |
AK139923.1 | - | 7 - 8 | 1345 | 109 | |
BC032277.1 | - | 8 - 9 | 1327 | 109 | |
L76944.1 | - | 8 - 9 | 1028 | 109 | |
L76946.1 | - | 9 - 10 | 1374 | 109 | |
L76947.1 | - | 8 - 9 | 861 | 109 |