The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004154.2 | NP_001004154.1 | 1 - 2 | 619 | 99 |
GenBank mRNA | BC069180.1 | - | 1 - 2 | 577 | 99 |
BC070407.1 | - | 1 - 2 | 596 | 99 |